The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297575.1 | NP_001284504.1 | 3 - 4 | 542 | 67 |
NM_014625.3 | NP_055440.1 | 3 - 4 | 542 | 67 | |
XM_017002299.1 | XP_016857788.1 | 3 - 4 | 577 | 67 | |
GenBank mRNA | AJ279254.1 | - | 3 - 4 | 523 | 67 |
BC029141.1 | - | 3 - 4 | 542 | 67 |