Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM16
Entrez Gene ID
63976
Gene Name
PR domain 16
Gene Aliases
CMD1LL, LVNC8, MEL1, PFM13
Chromosome Location
Chr.1: 3069025 - 3438621 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022114.3 NP_071397.3 4 - 5 655 68
NM_199454.2 NP_955533.2 4 - 5 655 68
XM_005244772.4 XP_005244829.1 4 - 5 807 68
XM_005244773.4 XP_005244830.1 4 - 5 811 68
XM_005244774.4 XP_005244831.1 4 - 5 810 68
XM_006710814.3 XP_006710877.1 4 - 5 808 68
XM_011541945.2 XP_011540247.1 3 - 4 428 68
XM_017002050.1 XP_016857539.1 4 - 5 658 68
GenBank mRNA AB051462.2 - 4 - 5 605 68
AB078876.1 - 4 - 5 617 68
AK309078.1 - 3 - 4 351 68
BC161614.1 - 4 - 5 605 68
BQ897896.1 - 4 - 5 656 68

View Full Product Details