Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM16
Entrez Gene ID
63976
Gene Name
PR domain 16
Gene Aliases
CMD1LL, LVNC8, MEL1, PFM13
Chromosome Location
Chr.1: 3069025 - 3438621 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022114.3 NP_071397.3 5 - 6 756 79
NM_199454.2 NP_955533.2 5 - 6 756 79
XM_005244772.4 XP_005244829.1 5 - 6 908 79
XM_005244773.4 XP_005244830.1 5 - 6 912 79
XM_005244774.4 XP_005244831.1 5 - 6 911 79
XM_006710814.3 XP_006710877.1 5 - 6 909 79
XM_011541945.2 XP_011540247.1 4 - 5 529 79
XM_017002050.1 XP_016857539.1 5 - 6 759 79
GenBank mRNA AB051462.2 - 5 - 6 706 79
AB078876.1 - 5 - 6 718 79
AF294278.1 - 5 - 6 740 79
AK309078.1 - 4 - 5 452 79
BC161614.1 - 5 - 6 706 79

View Full Product Details