The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022114.3 | NP_071397.3 | 11 - 12 | 2941 | 130 |
NM_199454.2 | NP_955533.2 | 11 - 12 | 2941 | 130 | |
XM_005244772.4 | XP_005244829.1 | 11 - 12 | 3093 | 130 | |
XM_005244773.4 | XP_005244830.1 | 11 - 12 | 3097 | 130 | |
XM_005244774.4 | XP_005244831.1 | 11 - 12 | 3096 | 130 | |
XM_006710814.3 | XP_006710877.1 | 11 - 12 | 3091 | 130 | |
XM_011541945.2 | XP_011540247.1 | 10 - 11 | 2714 | 130 | |
XM_017002050.1 | XP_016857539.1 | 11 - 12 | 2941 | 130 | |
GenBank mRNA | AB051462.2 | - | 11 - 12 | 2891 | 130 |
AB078876.1 | - | 11 - 12 | 2903 | 130 | |
AF294278.1 | - | 11 - 12 | 2922 | 130 | |
AK309078.1 | - | 10 - 11 | 2637 | 130 | |
BC161614.1 | - | 11 - 12 | 2891 | 130 |