The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173620.2 | NP_775891.2 | 10 - 11 | 1667 | 77 |
XM_005256346.3 | XP_005256403.1 | 9 - 10 | 1255 | 77 | |
XM_005256347.4 | XP_005256404.1 | 11 - 12 | 1302 | 77 | |
XM_006722280.3 | XP_006722343.1 | 9 - 10 | 1230 | 77 | |
XM_011523560.2 | XP_011521862.1 | 8 - 9 | 1198 | 77 | |
XM_011523561.2 | XP_011521863.1 | 9 - 10 | 1236 | 77 | |
XM_017024485.1 | XP_016879974.1 | 10 - 11 | 1166 | 77 | |
GenBank mRNA | AK074405.1 | - | 10 - 11 | 1141 | 77 |
AK295197.1 | - | 7 - 8 | 1359 | 77 | |
AK316085.1 | - | 6 - 7 | 1351 | 77 | |
BC018205.1 | - | 10 - 11 | 1173 | 77 | |
BC035561.1 | - | 10 - 11 | 1348 | 77 | |
FM204887.1 | - | 10 - 11 | 1162 | 77 |