The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011709.2 | NP_001011709.2 | 7 - 8 | 912 | 108 |
XM_011539277.1 | XP_011537579.1 | 5 - 6 | 536 | 108 | |
XM_011539278.1 | XP_011537580.1 | 7 - 8 | 816 | 108 | |
XM_011539279.1 | XP_011537581.1 | 6 - 7 | 790 | 108 | |
GenBank mRNA | AL833418.1 | - | 7 - 8 | 961 | 108 |
BC117224.1 | - | 7 - 8 | 912 | 108 |