The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136195.1 | NP_001129667.1 | 5 - 6 | 686 | 60 |
NM_001136196.1 | NP_001129668.1 | 4 - 5 | 513 | 60 | |
NM_013433.4 | NP_038461.2 | 4 - 5 | 499 | 60 | |
GenBank mRNA | AB210011.1 | - | 3 - 4 | 1066 | 60 |
AF007748.1 | - | 5 - 6 | 619 | 60 | |
AF019039.1 | - | 4 - 5 | 452 | 60 | |
AK056183.1 | - | 4 - 5 | 1025 | 60 | |
BC018314.1 | - | 4 - 5 | 390 | 60 | |
BC072420.1 | - | 4 - 5 | 472 | 60 | |
DA100622.1 | - | 4 - 5 | 514 | 60 |