Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD2
Entrez Gene ID
143162
Gene Name
FERM and PDZ domain containing 2
Gene Aliases
PDZD5C, PDZK4, PDZK5C
Chromosome Location
Chr.10: 48156559 - 48274898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018071.3 NP_001018081.3 10 - 11 1501 92
NM_001318191.1 NP_001305120.1 8 - 9 1145 92
XM_011539327.2 XP_011537629.1 10 - 11 1299 92
GenBank mRNA AK123038.1 - 8 - 9 92
AK303923.1 - 10 - 11 1199 92
CR749241.1 - 10 - 11 1473 92

View Full Product Details