Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD2
Entrez Gene ID
143162
Gene Name
FERM and PDZ domain containing 2
Gene Aliases
PDZD5C, PDZK4, PDZK5C
Chromosome Location
Chr.10: 48156559 - 48274898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018071.3 NP_001018081.3 14 - 15 2119 78
NM_001318191.1 NP_001305120.1 12 - 13 1760 78
XM_011539327.2 XP_011537629.1 14 - 15 1917 78
GenBank mRNA AK123038.1 - 12 - 13 1760 78
AK131386.1 - 12 - 13 1605 78
AK303923.1 - 14 - 15 1817 78

View Full Product Details