The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042704.1 | NP_001036169.1 | 10 - 11 | 1046 | 93 |
NM_001257139.1 | NP_001244068.1 | 9 - 10 | 917 | 93 | |
NM_004070.3 | NP_004061.3 | 10 - 11 | 1046 | 93 | |
GenBank mRNA | AK225550.1 | - | 10 - 11 | 1001 | 93 |
AK290814.1 | - | 10 - 11 | 1001 | 93 | |
AK298285.1 | - | 9 - 10 | 896 | 93 | |
BC048282.1 | - | 9 - 10 | 971 | 93 | |
BC053869.1 | - | 11 - 12 | 1094 | 93 | |
Z30643.1 | - | 9 - 10 | 988 | 93 |