The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170692.1 | NP_001164163.1 | 2 - 3 | 722 | 147 |
NM_001170693.1 | NP_001164164.1 | 2 - 3 | 722 | 147 | |
GenBank mRNA | AK097760.1 | - | 2 - 3 | 421 | 147 |
BC026188.1 | - | 2 - 3 | 400 | 147 | |
BC026194.2 | - | 2 - 3 | 407 | 147 | |
BC033545.1 | - | 2 - 3 | 407 | 147 |