The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153225.3 | NP_694957.3 | 3 - 4 | 701 | 78 |
XM_017013145.1 | XP_016868634.1 | 3 - 4 | 946 | 78 | |
GenBank mRNA | AK292970.1 | - | 3 - 4 | 601 | 78 |
AY040546.1 | - | 3 - 4 | 599 | 78 | |
BC042877.1 | - | 3 - 4 | 701 | 78 |