Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBM22
Entrez Gene ID
55696
Gene Name
RNA binding motif protein 22
Gene Aliases
Cwc2, ZC3H16, fSAP47
Chromosome Location
Chr.5: 150690790 - 150701107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018047.2 NP_060517.1 7 - 8 863 132
GenBank mRNA AK001152.1 - 7 - 8 818 132
AK297019.1 - 6 - 7 632 132
AL136933.1 - 7 - 8 792 132
BC003402.1 - 7 - 8 796 132

View Full Product Details