The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144888.1 | NP_001138360.1 | 4 - 5 | 387 | 65 |
NM_006340.2 | NP_006331.1 | 4 - 5 | 387 | 65 | |
NM_017450.2 | NP_059344.1 | 4 - 5 | 387 | 65 | |
NM_017451.2 | NP_059345.1 | 4 - 5 | 387 | 65 | |
XM_005256943.1 | XP_005257000.1 | 5 - 6 | 486 | 65 | |
XM_005256944.1 | XP_005257001.1 | 5 - 6 | 486 | 65 | |
XM_005256945.1 | XP_005257002.1 | 5 - 6 | 486 | 65 | |
XM_005256948.3 | XP_005257005.1 | 4 - 5 | 387 | 65 | |
XM_006721635.1 | XP_006721698.1 | 5 - 6 | 486 | 65 | |
XM_006721636.1 | XP_006721699.1 | 5 - 6 | 486 | 65 | |
XM_006721637.2 | XP_006721700.1 | 4 - 5 | 387 | 65 | |
XM_011524193.1 | XP_011522495.1 | 5 - 6 | 486 | 65 | |
XM_011524194.1 | XP_011522496.1 | 5 - 6 | 486 | 65 | |
XM_017024017.1 | XP_016879506.1 | 3 - 4 | 1144 | 65 | |
XM_017024018.1 | XP_016879507.1 | 2 - 3 | 606 | 65 | |
GenBank mRNA | AB015019.1 | - | 4 - 5 | 372 | 65 |
AB015020.1 | - | 4 - 5 | 372 | 65 | |
AB017119.1 | - | 4 - 5 | 372 | 65 | |
AB017120.1 | - | 4 - 5 | 372 | 65 | |
AK023688.1 | - | 5 - 6 | 482 | 65 | |
AK225612.1 | - | 4 - 5 | 384 | 65 | |
AK298629.1 | - | 2 - 3 | 222 | 65 | |
BC002495.2 | - | 4 - 5 | 362 | 65 | |
BC014020.2 | - | 4 - 5 | 374 | 65 | |
BC032559.1 | - | 4 - 5 | 385 | 65 | |
CB153324.1 | - | 4 - 5 | 412 | 65 | |
U70669.1 | - | 4 - 5 | 372 | 65 |