The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002741.3 | NP_002732.3 | 9 - 10 | 1589 | 88 |
NM_213560.1 | NP_998725.1 | 9 - 10 | 1456 | 88 | |
XM_011528128.1 | XP_011526430.1 | 9 - 10 | 1472 | 88 | |
GenBank mRNA | AK123007.1 | - | 9 - 10 | 1470 | 88 |
AK292130.1 | - | 9 - 10 | 1453 | 88 | |
AK313886.1 | - | 9 - 10 | 1429 | 88 | |
BC018575.1 | - | 3 - 4 | 307 | 88 | |
BC040061.1 | - | 9 - 10 | 1586 | 88 | |
BC094766.1 | - | 9 - 10 | 1620 | 88 | |
D26181.1 | - | 9 - 10 | 1457 | 88 | |
S75546.1 | - | 9 - 10 | 1505 | 88 | |
U33053.1 | - | 9 - 10 | 1505 | 88 |