The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000824.4 | NP_000815.1 | 9 - 10 | 1400 | 159 |
NM_001166060.1 | NP_001159532.1 | 9 - 10 | 1383 | 159 | |
XM_017008034.1 | XP_016863523.1 | 8 - 9 | 1194 | 159 | |
GenBank mRNA | AF094754.1 | - | 9 - 10 | 1363 | 159 |
AF094755.1 | - | 9 - 10 | 1363 | 159 | |
AK290617.1 | - | 9 - 10 | 1334 | 159 | |
BC032635.1 | - | 9 - 10 | 1339 | 159 | |
U33267.1 | - | 9 - 10 | 1274 | 159 |