Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST18
Entrez Gene ID
9705
Gene Name
ST18, C2H2C-type zinc finger
Gene Aliases
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location
Chr.8: 52110832 - 52409879 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014682.2 NP_055497.1 17 - 18 2988 85
XM_006716487.1 XP_006716550.1 17 - 18 2755 85
XM_011517629.1 XP_011515931.1 18 - 19 2911 85
XM_011517631.1 XP_011515933.1 19 - 20 2952 85
XM_011517632.1 XP_011515934.1 17 - 18 2766 85
XM_011517633.1 XP_011515935.1 17 - 18 2759 85
XM_011517634.1 XP_011515936.1 17 - 18 2955 85
XM_011517635.1 XP_011515937.1 17 - 18 2959 85
XM_011517636.2 XP_011515938.1 16 - 17 3036 85
XM_011517637.1 XP_011515939.1 15 - 16 2493 85
XM_011517638.2 XP_011515940.1 14 - 15 2534 85
XM_011517641.1 XP_011515943.1 19 - 20 2999 85
XM_011517642.1 XP_011515944.1 17 - 18 2665 85
XM_017014047.1 XP_016869536.1 17 - 18 3115 85
XM_017014048.1 XP_016869537.1 17 - 18 3119 85
XM_017014049.1 XP_016869538.1 18 - 19 3165 85
XM_017014050.1 XP_016869539.1 17 - 18 2952 85
XM_017014051.1 XP_016869540.1 16 - 17 2959 85
XM_017014052.1 XP_016869541.1 18 - 19 3169 85
XM_017014053.1 XP_016869542.1 18 - 19 3156 85
XM_017014054.1 XP_016869543.1 16 - 17 3205 85
XM_017014055.1 XP_016869544.1 16 - 17 2796 85
XM_017014056.1 XP_016869545.1 19 - 20 3206 85
XM_017014057.1 XP_016869546.1 17 - 18 3009 85
XM_017014058.1 XP_016869547.1 17 - 18 2770 85
XM_017014059.1 XP_016869548.1 17 - 18 2956 85
XM_017014060.1 XP_016869549.1 18 - 19 2993 85
XM_017014061.1 XP_016869550.1 16 - 17 3128 85
XM_017014062.1 XP_016869551.1 15 - 16 2880 85
XM_017014063.1 XP_016869552.1 16 - 17 3426 85
XM_017014064.1 XP_016869553.1 16 - 17 3040 85
XM_017014065.1 XP_016869554.1 18 - 19 3160 85
XM_017014066.1 XP_016869555.1 16 - 17 2799 85
XM_017014067.1 XP_016869556.1 16 - 17 3209 85
XM_017014068.1 XP_016869557.1 17 - 18 3000 85
XM_017014069.1 XP_016869558.1 14 - 15 2697 85
XM_017014070.1 XP_016869559.1 13 - 14 2618 85
XM_017014071.1 XP_016869560.1 14 - 15 2866 85
XM_017014072.1 XP_016869561.1 18 - 19 3030 85
XM_017014073.1 XP_016869562.1 18 - 19 2668 85
XM_017014074.1 XP_016869563.1 17 - 18 2951 85
XM_017014075.1 XP_016869564.1 18 - 19 3202 85
XM_017014076.1 XP_016869565.1 18 - 19 3206 85
XM_017014077.1 XP_016869566.1 18 - 19 2843 85
XM_017014078.1 XP_016869567.1 17 - 18 3123 85
XM_017014079.1 XP_016869568.1 17 - 18 3514 85
XM_017014080.1 XP_016869569.1 15 - 16 2755 85
XM_017014081.1 XP_016869570.1 16 - 17 2876 85
XM_017014082.1 XP_016869571.1 16 - 17 2711 85
XM_017014083.1 XP_016869572.1 15 - 16 2551 85
XM_017014084.1 XP_016869573.1 13 - 14 2289 85
GenBank mRNA AB011107.1 - 17 - 18 2852 85
BC117147.1 - 12 - 13 2245 85
BC117149.1 - 12 - 13 2245 85
BC143872.1 - 12 - 13 2240 85
BC143873.1 - 13 - 14 2333 85

View Full Product Details