Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CDKN2A
- Entrez Gene ID
-
1029
- Gene Name
- cyclin dependent kinase inhibitor 2A
- Gene Aliases
-
ARF, CDK4I, CDKN2, CMM2, INK4, INK4A, MLM, MTS-1, MTS1, P14, P14ARF, P16, P16-INK4A, P16INK4, P16INK4A, P19, P19ARF, TP16
- Chromosome Location
-
Chr.9: 21967752 - 21995043 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058197.4 |
NP_478104.2 |
1 - 2 |
416 |
101 |
View Full Product Details