The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003178.5 | NP_003169.2 | 4 - 5 | 875 | 100 |
NM_133625.4 | NP_598328.1 | 4 - 5 | 875 | 100 | |
XM_006713311.3 | XP_006713374.1 | 4 - 5 | 869 | 100 | |
GenBank mRNA | AB209088.1 | - | 5 - 6 | 793 | 100 |
AF077671.1 | - | 4 - 5 | 704 | 100 | |
AF077737.1 | - | 4 - 5 | 704 | 100 | |
AK094464.1 | - | 5 - 6 | 643 | 100 | |
AK295431.1 | - | 1 - 2 | 91 | 100 | |
BC051307.1 | - | 4 - 5 | 833 | 100 | |
U40215.1 | - | 4 - 5 | 739 | 100 |