Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLE
Entrez Gene ID
5426
Gene Name
DNA polymerase epsilon, catalytic subunit
Gene Aliases
CRCS12, FILS, POLE1
Chromosome Location
Chr.12: 132623762 - 132687524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006231.3 NP_006222.2 41 - 42 5889 86
XM_011534795.2 XP_011533097.1 41 - 42 5884 86
XM_011534797.2 XP_011533099.1 33 - 34 4853 86
XM_011534798.1 XP_011533100.1 29 - 30 4405 86
XM_011534802.2 XP_011533104.1 17 - 18 2706 86
GenBank mRNA AK093003.1 - 3 - 4 910 86
BC007599.1 - 7 - 8 1215 86
BC021559.1 - 2 - 3 194 86
BC087613.1 - 7 - 8 1215 86
BC144561.1 - 41 - 42 6396 86
BX647647.1 - 15 - 16 3334 86
L09561.1 - 41 - 42 5724 86
S60080.1 - 40 - 41 5606 86
U49356.1 - 41 - 42 5677 86

View Full Product Details