The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323517.1 | NP_001310446.1 | 2 - 3 | 282 | 61 |
NM_001323518.1 | NP_001310447.1 | 2 - 3 | 134 | 61 | |
NM_006744.3 | NP_006735.2 | 2 - 3 | 196 | 61 | |
GenBank mRNA | BC020633.1 | - | 2 - 3 | 196 | 61 |
BG565176.1 | - | 2 - 3 | 135 | 61 | |
X00129.1 | - | 2 - 3 | 157 | 61 |