Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF21A
Entrez Gene ID
55605
Gene Name
kinesin family member 21A
Gene Aliases
CFEOM1, FEOM1, FEOM3A
Chromosome Location
Chr.12: 39293228 - 39443414 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173463.1 NP_001166934.1 10 - 11 1885 106
NM_001173464.1 NP_001166935.1 10 - 11 1885 106
NM_001173465.1 NP_001166936.1 10 - 11 1885 106
NM_017641.3 NP_060111.2 10 - 11 1885 106
XM_005269007.2 XP_005269064.1 10 - 11 1909 106
XM_005269008.2 XP_005269065.1 10 - 11 1909 106
XM_005269009.2 XP_005269066.1 10 - 11 1909 106
XM_005269010.2 XP_005269067.1 10 - 11 1909 106
XM_005269011.2 XP_005269068.1 10 - 11 1909 106
XM_005269012.2 XP_005269069.1 10 - 11 1909 106
XM_005269013.2 XP_005269070.1 10 - 11 1909 106
XM_005269014.2 XP_005269071.1 10 - 11 1909 106
XM_006719493.2 XP_006719556.1 10 - 11 1909 106
XM_006719494.2 XP_006719557.1 10 - 11 1909 106
XM_011538556.2 XP_011536858.1 10 - 11 1909 106
XM_017019607.1 XP_016875096.1 10 - 11 1909 106
XM_017019608.1 XP_016875097.1 10 - 11 1909 106
XM_017019609.1 XP_016875098.1 10 - 11 1909 106
XM_017019610.1 XP_016875099.1 10 - 11 1909 106
XM_017019611.1 XP_016875100.1 10 - 11 1909 106
GenBank mRNA AB051495.2 - 10 - 11 1454 106
AB290166.1 - 10 - 11 1885 106
AF155117.1 - 10 - 11 1614 106
AF450487.1 - 10 - 11 1614 106
AK000059.1 - 10 - 11 1627 106
AM177179.1 - 10 - 11 1465 106
AY368076.1 - 10 - 11 1481 106
BC041430.1 - 10 - 11 1605 106
BC136414.1 - 10 - 11 1554 106

View Full Product Details