Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF21A
Entrez Gene ID
55605
Gene Name
kinesin family member 21A
Gene Aliases
CFEOM1, FEOM1, FEOM3A
Chromosome Location
Chr.12: 39293228 - 39443414 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173463.1 NP_001166934.1 2 - 3 691 135
NM_001173464.1 NP_001166935.1 2 - 3 691 135
NM_001173465.1 NP_001166936.1 2 - 3 691 135
NM_017641.3 NP_060111.2 2 - 3 691 135
XM_005269007.2 XP_005269064.1 2 - 3 715 135
XM_005269008.2 XP_005269065.1 2 - 3 715 135
XM_005269009.2 XP_005269066.1 2 - 3 715 135
XM_005269010.2 XP_005269067.1 2 - 3 715 135
XM_005269011.2 XP_005269068.1 2 - 3 715 135
XM_005269012.2 XP_005269069.1 2 - 3 715 135
XM_005269013.2 XP_005269070.1 2 - 3 715 135
XM_005269014.2 XP_005269071.1 2 - 3 715 135
XM_006719493.2 XP_006719556.1 2 - 3 715 135
XM_006719494.2 XP_006719557.1 2 - 3 715 135
XM_011538556.2 XP_011536858.1 2 - 3 715 135
XM_017019607.1 XP_016875096.1 2 - 3 715 135
XM_017019608.1 XP_016875097.1 2 - 3 715 135
XM_017019609.1 XP_016875098.1 2 - 3 715 135
XM_017019610.1 XP_016875099.1 2 - 3 715 135
XM_017019611.1 XP_016875100.1 2 - 3 715 135
GenBank mRNA AB051495.2 - 2 - 3 260 135
AB290166.1 - 2 - 3 691 135
AF155117.1 - 2 - 3 417 135
AF450487.1 - 2 - 3 417 135
AK000059.1 - 2 - 3 433 135
AM177179.1 - 2 - 3 271 135
AY368076.1 - 2 - 3 287 135
BC041430.1 - 2 - 3 411 135
BC136414.1 - 2 - 3 360 135

View Full Product Details