Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF21A
Entrez Gene ID
55605
Gene Name
kinesin family member 21A
Gene Aliases
CFEOM1, FEOM1, FEOM3A
Chromosome Location
Chr.12: 39293228 - 39443414 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173463.1 NP_001166934.1 5 - 6 1155 106
NM_001173464.1 NP_001166935.1 5 - 6 1155 106
NM_001173465.1 NP_001166936.1 5 - 6 1155 106
NM_017641.3 NP_060111.2 5 - 6 1155 106
XM_005269007.2 XP_005269064.1 5 - 6 1179 106
XM_005269008.2 XP_005269065.1 5 - 6 1179 106
XM_005269009.2 XP_005269066.1 5 - 6 1179 106
XM_005269010.2 XP_005269067.1 5 - 6 1179 106
XM_005269011.2 XP_005269068.1 5 - 6 1179 106
XM_005269012.2 XP_005269069.1 5 - 6 1179 106
XM_005269013.2 XP_005269070.1 5 - 6 1179 106
XM_005269014.2 XP_005269071.1 5 - 6 1179 106
XM_006719493.2 XP_006719556.1 5 - 6 1179 106
XM_006719494.2 XP_006719557.1 5 - 6 1179 106
XM_011538556.2 XP_011536858.1 5 - 6 1179 106
XM_017019607.1 XP_016875096.1 5 - 6 1179 106
XM_017019608.1 XP_016875097.1 5 - 6 1179 106
XM_017019609.1 XP_016875098.1 5 - 6 1179 106
XM_017019610.1 XP_016875099.1 5 - 6 1179 106
XM_017019611.1 XP_016875100.1 5 - 6 1179 106
GenBank mRNA AB051495.2 - 5 - 6 724 106
AB290166.1 - 5 - 6 1155 106
AF155117.1 - 5 - 6 884 106
AF450487.1 - 5 - 6 884 106
AK000059.1 - 5 - 6 897 106
AM177179.1 - 5 - 6 735 106
AY368076.1 - 5 - 6 751 106
BC041430.1 - 5 - 6 875 106
BC136414.1 - 5 - 6 824 106

View Full Product Details