The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014966.3 | NP_055781.2 | 16 - 17 | 2950 | 61 |
NM_138615.2 | NP_619520.1 | 15 - 16 | 2921 | 61 | |
XM_006713033.1 | XP_006713096.1 | 14 - 15 | 2744 | 61 | |
XM_011533490.1 | XP_011531792.1 | 17 - 18 | 3176 | 61 | |
XM_011533491.1 | XP_011531793.1 | 18 - 19 | 3363 | 61 | |
XM_011533492.2 | XP_011531794.1 | 16 - 17 | 3195 | 61 | |
XM_011533493.1 | XP_011531795.1 | 16 - 17 | 3065 | 61 | |
XM_011533494.2 | XP_011531796.1 | 15 - 16 | 2885 | 61 | |
XM_011533495.1 | XP_011531797.1 | 15 - 16 | 2730 | 61 | |
XM_011533496.1 | XP_011531798.1 | 16 - 17 | 3082 | 61 | |
XM_011533497.2 | XP_011531799.1 | 16 - 17 | 2963 | 61 | |
XM_011533498.1 | XP_011531800.1 | 16 - 17 | 3033 | 61 | |
XM_017005914.1 | XP_016861403.1 | 16 - 17 | 2826 | 61 | |
XM_017005915.1 | XP_016861404.1 | 14 - 15 | 2701 | 61 | |
XM_017005916.1 | XP_016861405.1 | 14 - 15 | 2786 | 61 | |
XM_017005917.1 | XP_016861406.1 | 17 - 18 | 2992 | 61 | |
GenBank mRNA | AB020697.1 | - | 15 - 16 | 2638 | 61 |
AK291266.1 | - | 16 - 17 | 2725 | 61 | |
BC014237.2 | - | 3 - 4 | 416 | 61 | |
BC015029.1 | - | 16 - 17 | 2696 | 61 | |
BC020126.1 | - | 15 - 16 | 2669 | 61 | |
BC038417.1 | - | 16 - 17 | 2697 | 61 |