The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174100.1 | NP_001167571.1 | 5 - 6 | 490 | 59 |
NM_020418.3 | NP_065151.2 | 5 - 6 | 393 | 59 | |
NM_033008.2 | NP_127501.1 | 4 - 5 | 342 | 59 | |
NM_033010.2 | NP_127503.1 | 4 - 5 | 284 | 59 | |
XM_005265330.2 | XP_005265387.1 | 5 - 6 | 2297 | 59 | |
XM_005265331.2 | XP_005265388.1 | 5 - 6 | 416 | 59 | |
XM_005265332.2 | XP_005265389.1 | 5 - 6 | 2297 | 59 | |
XM_006713268.2 | XP_006713331.1 | 5 - 6 | 2105 | 59 | |
XM_006713269.1 | XP_006713332.1 | 6 - 7 | 561 | 59 | |
XM_006713270.1 | XP_006713333.1 | 4 - 5 | 333 | 59 | |
XM_006713271.2 | XP_006713334.1 | 5 - 6 | 2297 | 59 | |
XM_017006916.1 | XP_016862405.1 | 6 - 7 | 718 | 59 | |
XM_017006917.1 | XP_016862406.1 | 5 - 6 | 2105 | 59 | |
XM_017006918.1 | XP_016862407.1 | 4 - 5 | 1957 | 59 | |
XM_017006919.1 | XP_016862408.1 | 4 - 5 | 333 | 59 | |
XM_017006920.1 | XP_016862409.1 | 4 - 5 | 333 | 59 | |
GenBank mRNA | AF092441.1 | - | 4 - 5 | 256 | 59 |
AF176330.1 | - | 4 - 5 | 284 | 59 | |
AK023993.1 | - | 4 - 5 | 331 | 59 | |
AK225815.1 | - | 5 - 6 | 469 | 59 | |
BC003008.1 | - | 4 - 5 | 305 | 59 | |
BC004153.2 | - | 4 - 5 | 305 | 59 | |
BC017098.1 | - | 5 - 6 | 388 | 59 |