Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PCBP4
- Entrez Gene ID
-
57060
- Gene Name
- poly(rC) binding protein 4
- Gene Aliases
-
CBP, LIP4, MCG10
- Chromosome Location
-
Chr.3: 51957454 - 51967466 on Build GRCh38
View Full Product Details