The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033008.2 | NP_127501.1 | 1 - 2 | 139 | 135 |
XM_006713270.1 | XP_006713333.1 | 1 - 2 | 130 | 135 | |
XM_017006919.1 | XP_016862408.1 | 1 - 2 | 130 | 135 | |
XM_017006920.1 | XP_016862409.1 | 1 - 2 | 130 | 135 | |
GenBank mRNA | AK023993.1 | - | 1 - 2 | 128 | 135 |