The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145247.4 | NP_660290.3 | 1 - 2 | 94 | 145 |
GenBank mRNA | AK123954.1 | - | 1 - 2 | 1034 | 145 |
BE960638.1 | - | - | 93 | 144 |