Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRMP
Entrez Gene ID
4033
Gene Name
lymphoid restricted membrane protein
Gene Aliases
JAW1
Chromosome Location
Chr.12: 25052101 - 25108335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204126.1 NP_001191055.1 8 - 9 1330 108
NM_001204127.1 NP_001191056.1 8 - 9 1260 108
NM_006152.3 NP_006143.2 9 - 10 1411 108
XM_005253370.3 XP_005253427.1 10 - 11 1470 108
XM_005253372.2 XP_005253429.1 9 - 10 1471 108
XM_005253374.3 XP_005253431.1 7 - 8 1328 108
XM_006719076.2 XP_006719139.1 6 - 7 394 108
XM_011520667.1 XP_011518969.1 10 - 11 1619 108
XM_011520668.2 XP_011518970.1 6 - 7 657 108
XM_011520669.2 XP_011518971.1 5 - 6 539 108
XM_011520670.1 XP_011518972.1 5 - 6 310 108
XM_017019300.1 XP_016874789.1 9 - 10 1554 108
XM_017019301.1 XP_016874790.1 8 - 9 1263 108
XM_017019302.1 XP_016874791.1 9 - 10 1468 108
GenBank mRNA AK303103.1 - 5 - 6 663 108
AK303256.1 - 8 - 9 869 108
BC126417.1 - 9 - 10 918 108
BC144164.1 - 8 - 9 837 108
BC144165.1 - 9 - 10 918 108
BQ420905.1 - 8 - 9 628 108
U10485.1 - 9 - 10 1059 108

View Full Product Details