Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRMP
Entrez Gene ID
4033
Gene Name
lymphoid restricted membrane protein
Gene Aliases
JAW1
Chromosome Location
Chr.12: 25052101 - 25108335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204126.1 NP_001191055.1 9 - 10 1386 97
NM_001204127.1 NP_001191056.1 9 - 10 1316 97
NM_006152.3 NP_006143.2 10 - 11 1467 97
XM_005253370.3 XP_005253427.1 11 - 12 1526 97
XM_005253372.2 XP_005253429.1 10 - 11 1527 97
XM_005253374.3 XP_005253431.1 8 - 9 1384 97
XM_006719076.2 XP_006719139.1 7 - 8 450 97
XM_011520667.1 XP_011518969.1 11 - 12 1675 97
XM_011520668.2 XP_011518970.1 7 - 8 713 97
XM_011520669.2 XP_011518971.1 6 - 7 595 97
XM_011520670.1 XP_011518972.1 6 - 7 366 97
XM_017019300.1 XP_016874789.1 10 - 11 1610 97
XM_017019301.1 XP_016874790.1 9 - 10 1319 97
XM_017019302.1 XP_016874791.1 10 - 11 1524 97
GenBank mRNA AK303103.1 - 6 - 7 719 97
AK303256.1 - 9 - 10 925 97
BC126417.1 - 10 - 11 974 97
BC144164.1 - 9 - 10 893 97
BC144165.1 - 10 - 11 974 97
BQ420905.1 - 9 - 10 684 97
U10485.1 - 10 - 11 1115 97

View Full Product Details