The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204126.1 | NP_001191055.1 | 19 - 20 | 2259 | 88 |
NM_001204127.1 | NP_001191056.1 | 19 - 20 | 2189 | 88 | |
NM_006152.3 | NP_006143.2 | 20 - 21 | 2340 | 88 | |
XM_005253370.3 | XP_005253427.1 | 21 - 22 | 2399 | 88 | |
XM_005253372.2 | XP_005253429.1 | 20 - 21 | 2400 | 88 | |
XM_005253374.3 | XP_005253431.1 | 18 - 19 | 2257 | 88 | |
XM_006719076.2 | XP_006719139.1 | 17 - 18 | 1323 | 88 | |
XM_011520667.1 | XP_011518969.1 | 21 - 22 | 2548 | 88 | |
XM_011520668.2 | XP_011518970.1 | 17 - 18 | 1586 | 88 | |
XM_011520669.2 | XP_011518971.1 | 16 - 17 | 1468 | 88 | |
XM_011520670.1 | XP_011518972.1 | 16 - 17 | 1239 | 88 | |
XM_017019300.1 | XP_016874789.1 | 20 - 21 | 2483 | 88 | |
XM_017019301.1 | XP_016874790.1 | 19 - 20 | 2192 | 88 | |
XM_017019302.1 | XP_016874791.1 | 20 - 21 | 2397 | 88 | |
GenBank mRNA | AK303103.1 | - | 16 - 17 | 1592 | 88 |
AK303256.1 | - | 19 - 20 | 1798 | 88 | |
BC126417.1 | - | 20 - 21 | 1847 | 88 | |
BC144164.1 | - | 19 - 20 | 1766 | 88 | |
BC144165.1 | - | 19 - 20 | 1745 | 88 | |
BU679233.1 | - | 1 - 2 | 88 | ||
U10485.1 | - | 20 - 21 | 1988 | 88 |