The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204126.1 | NP_001191055.1 | 4 - 5 | 1032 | 67 |
NM_001204127.1 | NP_001191056.1 | 4 - 5 | 962 | 67 | |
NM_006152.3 | NP_006143.2 | 5 - 6 | 1113 | 67 | |
XM_005253370.3 | XP_005253427.1 | 6 - 7 | 1172 | 67 | |
XM_005253372.2 | XP_005253429.1 | 5 - 6 | 1173 | 67 | |
XM_005253374.3 | XP_005253431.1 | 3 - 4 | 1030 | 67 | |
XM_006719076.2 | XP_006719139.1 | 2 - 3 | 96 | 67 | |
XM_011520667.1 | XP_011518969.1 | 6 - 7 | 1321 | 67 | |
XM_017019300.1 | XP_016874789.1 | 5 - 6 | 1256 | 67 | |
XM_017019301.1 | XP_016874790.1 | 4 - 5 | 965 | 67 | |
XM_017019302.1 | XP_016874791.1 | 5 - 6 | 1170 | 67 | |
GenBank mRNA | AK303256.1 | - | 4 - 5 | 571 | 67 |
BC029391.1 | - | 6 - 7 | 858 | 67 | |
BC100277.1 | - | 5 - 6 | 1091 | 67 | |
BC126417.1 | - | 5 - 6 | 620 | 67 | |
BC144164.1 | - | 4 - 5 | 539 | 67 | |
BC144165.1 | - | 5 - 6 | 620 | 67 | |
BM466207.1 | - | 5 - 6 | 761 | 67 | |
BQ420905.1 | - | 4 - 5 | 330 | 67 | |
U10485.1 | - | 5 - 6 | 761 | 67 |