The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003619.3 | NP_003610.2 | 7 - 8 | 1538 | 81 |
XM_005263318.4 | XP_005263375.1 | 7 - 8 | 1811 | 81 | |
XM_011532387.2 | XP_011530689.1 | 7 - 8 | 1813 | 81 | |
GenBank mRNA | AJ001531.1 | - | 7 - 8 | 1531 | 81 |