The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000250.1 | NP_000241.1 | 2 - 3 | 424 | 95 |
GenBank mRNA | BC130476.1 | - | 2 - 3 | 298 | 95 |
J02694.1 | - | 2 - 3 | 395 | 95 | |
M19507.1 | - | 2 - 3 | 424 | 95 | |
S56200.1 | - | 2 - 3 | 247 | 95 | |
X04876.1 | - | 2 - 3 | 410 | 95 |