The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000250.1 | NP_000241.1 | 9 - 10 | 1795 | 109 |
GenBank mRNA | BC130476.1 | - | 9 - 10 | 1669 | 109 |
J02694.1 | - | 9 - 10 | 1766 | 109 | |
M19507.1 | - | 9 - 10 | 1795 | 109 | |
S56200.1 | - | 9 - 10 | 1618 | 109 | |
X04876.1 | - | 9 - 10 | 1781 | 109 |