Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCF25
Entrez Gene ID
22980
Gene Name
transcription factor 25
Gene Aliases
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049
Chromosome Location
Chr.16: 89873583 - 89911384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014972.2 NP_055787.1 14 - 15 1709 58
XM_005256297.2 XP_005256354.1 14 - 15 1703 58
XM_005256298.2 XP_005256355.1 14 - 15 1703 58
XM_005256299.2 XP_005256356.1 14 - 15 1658 58
XM_011522956.2 XP_011521258.1 14 - 15 1718 58
XM_011522957.2 XP_011521259.1 14 - 15 1718 58
XM_011522958.2 XP_011521260.1 14 - 15 1673 58
XM_011522959.2 XP_011521261.1 13 - 14 1652 58
XM_011522960.2 XP_011521262.1 14 - 15 1718 58
XM_011522961.2 XP_011521263.1 15 - 16 1887 58
XM_017023047.1 XP_016878536.1 13 - 14 1637 58
XM_017023048.1 XP_016878537.1 14 - 15 1658 58
XM_017023049.1 XP_016878538.1 14 - 15 1673 58
XM_017023050.1 XP_016878539.1 14 - 15 1658 58
XM_017023051.1 XP_016878540.1 13 - 14 1637 58
XM_017023052.1 XP_016878541.1 13 - 14 1592 58
XM_017023053.1 XP_016878542.1 15 - 16 2325 58
XM_017023054.1 XP_016878543.1 15 - 16 1872 58
XM_017023055.1 XP_016878544.1 15 - 16 1827 58
XM_017023056.1 XP_016878545.1 15 - 16 1879 58
XM_017023057.1 XP_016878546.1 15 - 16 2310 58
XM_017023058.1 XP_016878547.1 14 - 15 2244 58
GenBank mRNA AB028972.1 - 12 - 13 1250 58
AF130092.1 - 11 - 12 1211 58
AF322111.1 - 14 - 15 1723 58
AK095785.1 - 1 - 2 1146 58
BC000959.1 - 14 - 15 1632 58
BC009349.2 - 14 - 15 1654 58
BC011884.2 - 14 - 15 1683 58
DQ321703.1 - 14 - 15 1723 58

View Full Product Details