Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRPF40A
Entrez Gene ID
55660
Gene Name
pre-mRNA processing factor 40 homolog A
Gene Aliases
FBP-11, FBP11, FLAF1, FNBP3, HIP-10, HIP10, HYPA, NY-REN-6, Prp40
Chromosome Location
Chr.2: 152651593 - 152718007 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017892.3 NP_060362.3 20 - 21 2251 75
XM_005246676.4 XP_005246733.3 20 - 21 2771 75
XM_005246677.4 XP_005246734.2 20 - 21 2259 75
XM_011511446.2 XP_011509748.1 19 - 20 2729 75
XM_011511447.2 XP_011509749.1 20 - 21 2783 75
XM_011511448.2 XP_011509750.1 19 - 20 2205 75
XM_011511449.2 XP_011509751.1 22 - 23 2613 75
XM_017004453.1 XP_016859942.1 20 - 21 2783 75
XM_017004454.1 XP_016859943.1 19 - 20 2717 75
XM_017004455.1 XP_016859944.1 19 - 20 2193 75
XM_017004456.1 XP_016859945.1 21 - 22 2449 75
XM_017004457.1 XP_016859946.1 22 - 23 2600 75
XM_017004458.1 XP_016859947.1 20 - 21 2359 75
XM_017004459.1 XP_016859948.1 20 - 21 2468 75
XM_017004460.1 XP_016859949.1 20 - 21 2384 75
XM_017004461.1 XP_016859950.1 20 - 21 2456 75
GenBank mRNA AF151059.1 - 4 - 5 383 75
AF155096.1 - 6 - 7 688 75
AK000592.1 - 2 - 3 183 75
AK021621.1 - 5 - 6 1339 75
AK298547.1 - 17 - 18 2100 75
AK315187.1 - 2 - 3 215 75
BC027178.1 - 1 - 2 85 75

View Full Product Details