The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278352.1 | NP_001265281.1 | 17 - 18 | 2186 | 127 |
NM_002197.2 | NP_002188.1 | 16 - 17 | 2114 | 127 | |
XM_005251476.1 | XP_005251533.1 | 17 - 18 | 2124 | 127 | |
XM_011517888.1 | XP_011516190.1 | 16 - 17 | 2036 | 127 | |
GenBank mRNA | AB209480.1 | - | 16 - 17 | 2069 | 127 |
AF261088.1 | - | 17 - 18 | 2165 | 127 | |
AK308752.1 | - | 15 - 16 | 2080 | 127 | |
BC018103.1 | - | 15 - 16 | 1996 | 127 | |
EU794627.1 | - | 15 - 16 | 1996 | 127 | |
M58510.1 | - | 14 - 15 | 1742 | 127 | |
Z11559.1 | - | 16 - 17 | 2066 | 127 |