Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACO1
Entrez Gene ID
48
Gene Name
aconitase 1
Gene Aliases
ACONS, HEL60, IREB1, IREBP, IREBP1, IRP1
Chromosome Location
Chr.9: 32384603 - 32450836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278352.1 NP_001265281.1 18 - 19 2331 64
NM_002197.2 NP_002188.1 17 - 18 2259 64
XM_005251476.1 XP_005251533.1 18 - 19 2269 64
XM_011517888.1 XP_011516190.1 17 - 18 2181 64
GenBank mRNA AF261088.1 - 18 - 19 2310 64
BC018103.1 - 16 - 17 2141 64
EU794627.1 - 16 - 17 2141 64
M58510.1 - 15 - 16 1887 64
Z11559.1 - 17 - 18 2211 64

View Full Product Details