The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278352.1 | NP_001265281.1 | 3 - 4 | 325 | 109 |
NM_002197.2 | NP_002188.1 | 2 - 3 | 253 | 109 | |
XM_005251476.1 | XP_005251533.1 | 3 - 4 | 263 | 109 | |
XM_011517888.1 | XP_011516190.1 | 2 - 3 | 175 | 109 | |
GenBank mRNA | AB209480.1 | - | 2 - 3 | 208 | 109 |
AK057904.1 | - | 2 - 3 | 221 | 109 | |
AK308752.1 | - | 2 - 3 | 205 | 109 | |
BC018103.1 | - | 1 - 2 | 135 | 109 | |
BP287929.1 | - | 3 - 4 | 236 | 109 | |
DA103947.1 | - | 3 - 4 | 272 | 109 | |
DA409113.1 | - | 2 - 3 | 253 | 109 | |
DA780353.1 | - | 2 - 3 | 253 | 109 | |
EU794627.1 | - | 1 - 2 | 135 | 109 | |
Z11559.1 | - | 2 - 3 | 205 | 109 |