Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TYROBP
Entrez Gene ID
7305
Gene Name
TYRO protein tyrosine kinase binding protein
Gene Aliases
DAP12, KARAP, PLOSL
Chromosome Location
Chr.19: 35904401 - 35908309 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003332.3 NP_003323.1 1 - 2 135 127
NM_198125.2 NP_937758.1 1 - 2 135 127
GenBank mRNA AF019562.1 - 1 - 2 121 127
AK290385.1 - 1 - 2 121 127
BC011175.1 - 1 - 2 110 127
BP302545.1 - 1 - 2 135 127

View Full Product Details