The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136053.2 | NP_001129525.1 | 10 - 11 | 1224 | 72 |
NM_001142646.2 | NP_001136118.1 | 8 - 9 | 1060 | 72 | |
NR_073377.1 | 9 - 10 | 1163 | 72 | ||
XM_006713495.3 | XP_006713558.1 | 11 - 12 | 1405 | 72 | |
XM_006713496.2 | XP_006713559.1 | 10 - 11 | 1082 | 72 | |
XM_006713497.3 | XP_006713560.1 | 11 - 12 | 1449 | 72 | |
XM_011512422.1 | XP_011510724.1 | 10 - 11 | 998 | 72 | |
XM_017005707.1 | XP_016861196.1 | 11 - 12 | 1188 | 72 | |
XM_017005708.1 | XP_016861197.1 | 11 - 12 | 1218 | 72 | |
XM_017005709.1 | XP_016861198.1 | 10 - 11 | 1258 | 72 | |
XM_017005710.1 | XP_016861199.1 | 10 - 11 | 1097 | 72 | |
GenBank mRNA | AB037108.1 | - | 9 - 10 | 1148 | 72 |
AF289568.1 | - | 8 - 9 | 1007 | 72 | |
BC017058.1 | - | 9 - 10 | 932 | 72 | |
BC050711.2 | - | 10 - 11 | 1118 | 72 |