Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPRA1
Entrez Gene ID
131601
Gene Name
transmembrane protein adipocyte associated 1
Gene Aliases
GPR175, TMEM227, TPRA40
Chromosome Location
Chr.3: 127573064 - 127598234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136053.2 NP_001129525.1 6 - 7 862 58
NM_001142646.2 NP_001136118.1 6 - 7 862 58
NR_073377.1 6 - 7 862 58
XM_006713495.3 XP_006713558.1 7 - 8 1043 58
XM_006713496.2 XP_006713559.1 6 - 7 720 58
XM_006713497.3 XP_006713560.1 7 - 8 1087 58
XM_011512422.1 XP_011510724.1 6 - 7 636 58
XM_017005707.1 XP_016861196.1 7 - 8 826 58
XM_017005708.1 XP_016861197.1 7 - 8 856 58
GenBank mRNA AB037108.1 - 5 - 6 786 58
AF289568.1 - 6 - 7 809 58
BC017058.1 - 6 - 7 631 58
BC050711.2 - 6 - 7 756 58
CD580484.2 - 6 - 7 597 58

View Full Product Details