The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136053.2 | NP_001129525.1 | 7 - 8 | 976 | 93 |
XM_006713495.3 | XP_006713558.1 | 8 - 9 | 1157 | 93 | |
XM_006713496.2 | XP_006713559.1 | 7 - 8 | 834 | 93 | |
XM_006713497.3 | XP_006713560.1 | 8 - 9 | 1201 | 93 | |
XM_011512422.1 | XP_011510724.1 | 7 - 8 | 750 | 93 | |
XM_017005707.1 | XP_016861196.1 | 8 - 9 | 940 | 93 | |
XM_017005708.1 | XP_016861197.1 | 8 - 9 | 970 | 93 | |
GenBank mRNA | AB037108.1 | - | 6 - 7 | 900 | 93 |
BC050711.2 | - | 7 - 8 | 870 | 93 | |
CD580484.2 | - | 7 - 8 | 711 | 93 |