The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031478.5 | NP_113666.2 | 1 - 2 | 458 | 58 |
GenBank mRNA | AB593089.1 | - | 1 - 2 | 398 | 58 |
AB593090.1 | - | 1 - 2 | 396 | 58 | |
AF370365.1 | - | 1 - 2 | 410 | 58 | |
AL535138.3 | - | 1 - 2 | 466 | 58 | |
BC007892.2 | - | 1 - 2 | 176 | 58 | |
BQ185065.1 | - | 1 - 2 | 58 |