Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMC4
Entrez Gene ID
100272147
Gene Name
C-X9-C motif containing 4
Gene Aliases
C6.1B, MTCP1, MTCP1B, MTCP1NB, p8, p8MTCP1
Chromosome Location
Chr.X: 155061622 - 155071272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018024.2 NP_001018024.1 1 - 2 568 71
GenBank mRNA BC002600.1 - 1 - 2 530 71
BG707795.1 - 1 - 2 572 71
X64644.1 - 1 - 2 554 71

View Full Product Details