Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLNA
Entrez Gene ID
2316
Gene Name
filamin A
Gene Aliases
ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
Chromosome Location
Chr.X: 154348529 - 154374638 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110556.1 NP_001104026.1 15 - 16 2530 78
NM_001456.3 NP_001447.2 15 - 16 2530 78
XM_011531127.1 XP_011529429.1 15 - 16 2525 78
XM_011531128.1 XP_011529430.1 15 - 16 2525 78
XM_011531129.1 XP_011529431.1 15 - 16 2525 78
XM_011531130.1 XP_011529432.1 15 - 16 2525 78
XM_011531131.1 XP_011529433.1 14 - 15 2324 78
GenBank mRNA AB191259.1 - 14 - 15 2253 78
AB191260.1 - 14 - 15 2258 78
AB371574.1 - 14 - 15 2258 78
AB371575.1 - 14 - 15 2253 78
AB371576.1 - 14 - 15 2258 78
AB371577.1 - 14 - 15 2253 78
AB371578.1 - 14 - 15 2253 78
AB371579.1 - 14 - 15 2253 78
AB593010.1 - 14 - 15 2393 78
AK090427.2 - 14 - 15 2319 78
GU727643.1 - 15 - 16 2530 78
X53416.1 - 15 - 16 2453 78

View Full Product Details