The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110556.1 | NP_001104026.1 | 28 - 29 | 5003 | 98 |
NM_001456.3 | NP_001447.2 | 28 - 29 | 5003 | 98 | |
XM_011531127.1 | XP_011529429.1 | 28 - 29 | 4998 | 98 | |
XM_011531128.1 | XP_011529430.1 | 28 - 29 | 4998 | 98 | |
XM_011531129.1 | XP_011529431.1 | 28 - 29 | 4998 | 98 | |
XM_011531130.1 | XP_011529432.1 | 28 - 29 | 4998 | 98 | |
XM_011531131.1 | XP_011529433.1 | 27 - 28 | 4797 | 98 | |
GenBank mRNA | AB191259.1 | - | 27 - 28 | 4726 | 98 |
AB191260.1 | - | 27 - 28 | 4731 | 98 | |
AB371574.1 | - | 27 - 28 | 4731 | 98 | |
AB371575.1 | - | 27 - 28 | 4726 | 98 | |
AB371576.1 | - | 27 - 28 | 4731 | 98 | |
AB371577.1 | - | 27 - 28 | 4726 | 98 | |
AB371578.1 | - | 27 - 28 | 4726 | 98 | |
AB371579.1 | - | 27 - 28 | 4726 | 98 | |
AB593010.1 | - | 27 - 28 | 4866 | 98 | |
AK074048.1 | - | 7 - 8 | 1203 | 98 | |
AK090427.2 | - | 27 - 28 | 4792 | 98 | |
AL157419.1 | - | 5 - 6 | 767 | 98 | |
GU727643.1 | - | 28 - 29 | 5003 | 98 | |
X53416.1 | - | 28 - 29 | 4926 | 98 |