The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110556.1 | NP_001104026.1 | 3 - 4 | 869 | 137 |
NM_001456.3 | NP_001447.2 | 3 - 4 | 869 | 137 | |
XM_011531127.1 | XP_011529429.1 | 3 - 4 | 864 | 137 | |
XM_011531128.1 | XP_011529430.1 | 3 - 4 | 864 | 137 | |
XM_011531129.1 | XP_011529431.1 | 3 - 4 | 864 | 137 | |
XM_011531130.1 | XP_011529432.1 | 3 - 4 | 864 | 137 | |
XM_011531131.1 | XP_011529433.1 | 3 - 4 | 864 | 137 | |
GenBank mRNA | AB191259.1 | - | 2 - 3 | 592 | 137 |
AB191260.1 | - | 2 - 3 | 597 | 137 | |
AB371574.1 | - | 2 - 3 | 597 | 137 | |
AB371575.1 | - | 2 - 3 | 592 | 137 | |
AB371576.1 | - | 2 - 3 | 597 | 137 | |
AB371577.1 | - | 2 - 3 | 592 | 137 | |
AB371578.1 | - | 2 - 3 | 592 | 137 | |
AB371579.1 | - | 2 - 3 | 592 | 137 | |
AB593010.1 | - | 2 - 3 | 732 | 137 | |
AK090427.2 | - | 2 - 3 | 658 | 137 | |
GU727643.1 | - | 3 - 4 | 869 | 137 | |
X53416.1 | - | 3 - 4 | 792 | 137 |